gRNA Design Details
- Target DNA Sequence
- TGACAAGGGTTGCTGGCACA
- PAM Sequence
- AGG
- Target locus
-
Chr.2: 229591854 - 229591876 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
63.52/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- DNER
- Entrez Gene ID
-
92737
- Gene Name
- delta/notch like EGF repeat containing
- Gene Aliases
-
UNQ26, bet
- Chromosome Location
-
Chr.2: 229357629 - 229714570 on GRCh38
- UniGene ID
- Hs.234074
- Species
-
Human