gRNA Design Details
- Target DNA Sequence
- TCCATGACGTACACACGTTC
- PAM Sequence
- AGG
- Target locus
-
Chr.18: 21168758 - 21168780 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
87.59/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Garem
- Entrez Gene ID
-
381126
- Gene Name
- GRB2 associated, regulator of MAPK1
- Gene Aliases
-
C86169, Fam59a, Gm944, mKIAA4238
- Chromosome Location
-
Chr.18: 21127342 - 21300139 on GRCm38
- UniGene ID
- Mm.312276
- Species
-
Mouse