gRNA Design Details
- Target DNA Sequence
- CTATGTCCAGCTGGTGGTAC
- PAM Sequence
- AGG
- Target locus
-
Chr.15: 88816072 - 88816094 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
86.6/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Alg12
- Entrez Gene ID
-
223774
- Gene Name
- asparagine-linked glycosylation 12 (alpha-1,6-mannosyltransferase)
- Gene Aliases
-
ECM39
- Chromosome Location
-
Chr.15: 88805243 - 88819318 on GRCm38
- UniGene ID
- Mm.41512
- Species
-
Mouse