gRNA Design Details
- Target DNA Sequence
- GCTTGTTGGAGTGCTTGTTG
- PAM Sequence
- CGG
- Application
- Gene Knock-Out
Score:
88.05/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Gab3
- Entrez Gene ID
-
210710
- Gene Name
- growth factor receptor bound protein 2-associated protein 3
- Gene Aliases
-
5930433H21Rik
- Chromosome Location
-
Chr.X: 74988545 - 75084905 on GRCm38
- UniGene ID
- Mm.267056
- Species
-
Mouse