gRNA Design Details
- Target DNA Sequence
- GGTCTGCTTGATAGTGGTCA
- PAM Sequence
- TGG
- Target locus
-
Chr.5: 91362348 - 91362370 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
86.82/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Btc
- Entrez Gene ID
-
12223
- Gene Name
- betacellulin, epidermal growth factor family member
- Gene Aliases
-
Bcn
- Chromosome Location
-
Chr.5: 91357261 - 91402994 on GRCm38
- UniGene ID
- Mm.2024
- Species
-
Mouse