gRNA Design Details
- Target DNA Sequence
- GAAGTGGTCATTATGGACCT
- PAM Sequence
- GGG
- Target locus
-
Chr.15: 7254507 - 7254529 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
89.83/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Egflam
- Entrez Gene ID
-
268780
- Gene Name
- EGF-like, fibronectin type III and laminin G domains
- Gene Aliases
-
5930412K08, AU040377
- Chromosome Location
-
Chr.15: 7206120 - 7398746 on GRCm38
- UniGene ID
- Mm.203208
- Species
-
Mouse