gRNA Design Details
- Target DNA Sequence
- TGTCGTCTCCTGGGTATCGG
- PAM Sequence
- AGG
- Target locus
-
Chr.3: 108415133 - 108415155 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
90.59/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Celsr2
- Entrez Gene ID
-
53883
- Gene Name
- cadherin, EGF LAG seven-pass G-type receptor 2 (flamingo homolog, Drosophila)
- Gene Aliases
-
Adgrc2, EGFL2, Flamingo1, flamingo, mKIAA0279, mfmi1
- Chromosome Location
-
Chr.3: 108390848 - 108416026 on GRCm38
- UniGene ID
- Mm.39728
- Species
-
Mouse