gRNA Design Details
- Target DNA Sequence
- CGGTCTCGTAGAGAGCCTTG
- PAM Sequence
- AGG
- Target locus
-
Chr.18: 56708346 - 56708368 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
82.67/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Lmnb1
- Entrez Gene ID
-
16906
- Gene Name
- lamin B1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.18: 56707813 - 56753424 on GRCm38
- UniGene ID
- Mm.4105
- Species
-
Mouse