gRNA Design Details
- Target DNA Sequence
- GCGAAGTGGTTTGCTCGGGA
- PAM Sequence
- TGG
- Target locus
-
Chr.8: 80879571 - 80879593 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
93.18/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Gab1
- Entrez Gene ID
-
14388
- Gene Name
- growth factor receptor bound protein 2-associated protein 1
- Gene Aliases
-
AA408973, AW107238
- Chromosome Location
-
Chr.8: 80764431 - 80880755 on GRCm38
- UniGene ID
- Mm.277409
- Species
-
Mouse