gRNA Design Details
- Target DNA Sequence
- CCTGATTGAAGCCGCAGATC
- PAM Sequence
- TGG
- Target locus
-
Chr.7: 97223736 - 97223758 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
88.85/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Gab2
- Entrez Gene ID
-
14389
- Gene Name
- growth factor receptor bound protein 2-associated protein 2
- Gene Aliases
-
AI463667, D130058I17Rik, p97
- Chromosome Location
-
Chr.7: 97081751 - 97308951 on GRCm38
- UniGene ID
- Mm.42033
- Species
-
Mouse