gRNA Design Details
- Target DNA Sequence
- TCTCACCATAAACGCAACAG
- PAM Sequence
- TGG
- Target locus
-
Chr.11: 95891886 - 95891908 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
84.57/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- B4galnt2
- Entrez Gene ID
-
14422
- Gene Name
- beta-1,4-N-acetyl-galactosaminyl transferase 2
- Gene Aliases
-
AI593864, Dlb-1, Dlb1, Galg2, Galgt2, Ggm3, Igf2bp1
- Chromosome Location
-
Chr.11: 95863559 - 95914920 on GRCm38
- UniGene ID
- Mm.340702
- Species
-
Mouse