gRNA Design Details
- Target DNA Sequence
- ATAGGCACCGCCTGGAGTTC
- PAM Sequence
- TGG
- Target locus
-
Chr.17: 35201747 - 35201769 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
86.48/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Tnf
- Entrez Gene ID
-
21926
- Gene Name
- tumor necrosis factor
- Gene Aliases
-
DIF, TNF-a, TNF-alpha, TNFSF2, TNFalpha, Tnfa, Tnfsf1a
- Chromosome Location
-
Chr.17: 35199367 - 35202007 on GRCm38
- UniGene ID
- Mm.1293
- Species
-
Mouse