gRNA Design Details
- Target DNA Sequence
- AGAAGCGTCTCACTCCCGAA
- PAM Sequence
- GGG
- Target locus
-
Chr.5: 98254479 - 98254501 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
83.7/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Fgf5
- Entrez Gene ID
-
14176
- Gene Name
- fibroblast growth factor 5
- Gene Aliases
-
Fgf-5, HBGF-5, angora, go
- Chromosome Location
-
Chr.5: 98254184 - 98277033 on GRCm38
- UniGene ID
- Mm.5055
- Species
-
Mouse