gRNA Design Details
- Target DNA Sequence
- AAGGAAGTGAGACGGCACTT
- PAM Sequence
- CGG
- Target locus
-
Chr.22: 49910535 - 49910557 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
87.42/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- ALG12
- Entrez Gene ID
-
79087
- Gene Name
- ALG12, alpha-1,6-mannosyltransferase
- Gene Aliases
-
CDG1G, ECM39, PP14673, hALG12
- Chromosome Location
-
Chr.22: 49860163 - 49918469 on GRCh38
- UniGene ID
- Hs.526711
- Species
-
Human