Targeted Isoforms | Translated Protein | Targeted Exon(s) | |
---|---|---|---|
RefSeq | NM_001258374.1 | NP_001245303.1 | 2 |
NM_001258375.1 | NP_001245304.1 | 2 | |
NM_001258376.1 | NP_001245305.1 | 2 | |
NM_021235.2 | NP_067058.1 | 2 | |
NR_047665.1 | 2 | ||
NR_047666.1 | 2 | ||
XM_017027086.1 | XP_016882575.1 | 2 | |
XM_017027087.1 | XP_016882576.1 | 2 | |
XM_017027088.1 | XP_016882577.1 | 2 | |
XM_017027089.1 | XP_016882578.1 | 2 | |
XM_017027090.1 | XP_016882579.1 | 2 | |
XM_017027091.1 | XP_016882580.1 | 2 | |
XM_017027092.1 | XP_016882581.1 | 2 |