gRNA Design Details

Target DNA Sequence
GGCTTCATGTTGGCTCATGA
PAM Sequence
CGG
Target locus
Chr.11: 9079398 - 9079420 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 89.76/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
SCUBE2
Entrez Gene ID
57758
Gene Name
signal peptide, CUB domain and EGF like domain containing 2
Gene Aliases
CEGB1, CEGF1, CEGP1
Chromosome Location
Chr.11: 9019500 - 9091603 on GRCh38
UniGene ID
Hs.523468
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001170690.1 NP_001164161.1 3
NM_020974.2 NP_066025.2 3
XM_005253032.4 XP_005253089.2 3
XM_005253033.4 XP_005253090.2 3
XM_005253034.4 XP_005253091.2 3
XM_005253035.4 XP_005253092.2 3
XM_005253036.4 XP_005253093.2 3
XM_005253037.4 XP_005253094.2 3
XM_011520246.2 XP_011518548.1 3
XM_011520247.2 XP_011518549.1 3
XM_011520248.2 XP_011518550.1 3
XM_017018080.1 XP_016873569.1 3
XM_017018081.1 XP_016873570.1 3
XM_017018082.1 XP_016873571.1 3