gRNA Design Details
- Target DNA Sequence
- TTCCGTGGGAAAATTCCACG
- PAM Sequence
- TGG
- Target locus
-
Chr.17: 49133043 - 49133065 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
87.61/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- B4GALNT2
- Entrez Gene ID
-
124872
- Gene Name
- beta-1,4-N-acetyl-galactosaminyltransferase 2
- Gene Aliases
-
B4GALT, GALGT2
- Chromosome Location
-
Chr.17: 49120347 - 49170722 on GRCh38
- UniGene ID
- Hs.374679
- Species
-
Human