Targeted Isoforms | Translated Protein | Targeted Exon(s) | |
---|---|---|---|
RefSeq | NM_001159969.1 | NP_001153441.1 | 1 |
NM_001981.2 | NP_001972.1 | 13 | |
XM_005270618.2 | XP_005270675.1 | 12 | |
XM_017000615.1 | XP_016856104.1 | 13 | |
XM_017000616.1 | XP_016856105.1 | 13 | |
XM_017000617.1 | XP_016856106.1 | 13 | |
XM_017000618.1 | XP_016856107.1 | 12 | |
XM_017000619.1 | XP_016856108.1 | 14 | |
XM_017000620.1 | XP_016856109.1 | 13 |