gRNA Design Details
- Target DNA Sequence
- GCACTGGCTCCTTTATATCT
- PAM Sequence
- CGG
- Target locus
-
Chr.18: 32310276 - 32310298 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
88.87/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- GAREM1
- Entrez Gene ID
-
64762
- Gene Name
- GRB2 associated regulator of MAPK1 subtype 1
- Gene Aliases
-
C18orf11, FAM59A, GAREM, Gm944
- Chromosome Location
-
Chr.18: 32263521 - 32470518 on GRCh38
- UniGene ID
- Hs.444314
- Species
-
Human