gRNA Design Details
- Target DNA Sequence
- GCTGGCGGTGTACATCGACA
- PAM Sequence
- AGG
- Target locus
-
Chr.5: 126777634 - 126777656 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
88.98/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- LMNB1
- Entrez Gene ID
-
4001
- Gene Name
- lamin B1
- Gene Aliases
-
ADLD, LMN, LMN2, LMNB
- Chromosome Location
-
Chr.5: 126776623 - 126837020 on GRCh38
- UniGene ID
- Hs.89497
- Species
-
Human