gRNA Design Details
- Target DNA Sequence
- TTGGTGTTCGCTAAGATGAA
- PAM Sequence
- GGG
- Target locus
-
Chr.19: 4472378 - 4472400 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
86.73/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- HDGFRP2
- Entrez Gene ID
-
84717
- Gene Name
- hepatoma-derived growth factor-related protein 2
- Gene Aliases
-
HDGF-2, HDGF2, HRP-2
- Chromosome Location
-
Chr.19: 4472196 - 4502220 on GRCh38
- UniGene ID
- Hs.43071
- Species
-
Human