gRNA Design Details
- Target DNA Sequence
- CCACGATCAGGAAGGAGAAG
- PAM Sequence
- AGG
- Target locus
-
Chr.6: 31575843 - 31575865 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
86.38/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- TNF
- Entrez Gene ID
-
7124
- Gene Name
- tumor necrosis factor
- Gene Aliases
-
DIF, TNF-alpha, TNFA, TNFSF2, TNLG1F
- Chromosome Location
-
Chr.6: 31575567 - 31578336 on GRCh38
- UniGene ID
- Hs.241570
- Species
-
Human