gRNA Design Details

Target DNA Sequence
GTCCACTGTATTGGTGCAGG
PAM Sequence
TGG
Target locus
Chr.19: 6896437 - 6896459 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 89.4/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
ADGRE1
Entrez Gene ID
2015
Gene Name
adhesion G protein-coupled receptor E1
Gene Aliases
EMR1, TM7LN3
Chromosome Location
Chr.19: 6887549 - 6940453 on GRCh38
UniGene ID
Hs.2375
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001256254.1 NP_001243183.1 3