gRNA Design Details
- Target DNA Sequence
- AGGACTTGCGCTCTCGGAGA
- PAM Sequence
- AGG
- Target locus
-
Chr.11: 78250302 - 78250324 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
86.43/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- GAB2
- Entrez Gene ID
-
9846
- Gene Name
- GRB2 associated binding protein 2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.11: 78215290 - 78417822 on GRCh38
- UniGene ID
- Hs.429434
- Species
-
Human