Targeted Isoforms | Translated Protein | Targeted Exon(s) | |
---|---|---|---|
RefSeq | NM_001081573.2 | NP_001075042.1 | 1 |
NM_001282283.1 | NP_001269212.1 | 1 | |
NM_080612.3 | NP_542179.1 | 1 | |
XM_005274648.1 | XP_005274705.1 | 1 | |
XM_006724804.1 | XP_006724867.1 | 1 | |
XM_011531103.1 | XP_011529405.1 | 1 | |
XM_011531105.1 | XP_011529407.1 | 1 | |
XM_011531106.1 | XP_011529408.1 | 1 | |
XM_017029276.1 | XP_016884765.1 | 1 |