gRNA Design Details
- Target DNA Sequence
- GTGAAGTGGTCTGCTCCGGA
- PAM Sequence
- TGG
- Target locus
-
Chr.4: 143337199 - 143337221 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
88.33/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- GAB1
- Entrez Gene ID
-
2549
- Gene Name
- GRB2 associated binding protein 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.4: 143336830 - 143474565 on GRCh38
- UniGene ID
- Hs.618456
- Species
-
Human