gRNA Design Details

Target DNA Sequence
CTACTTGGTAGCAGACAGCG
PAM Sequence
AGG
Target locus
Chr.4: 143415686 - 143415708 on GRCh38
Strand
Forward
Application
Gene Knock-Out
Score: 86.79/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
GAB1
Entrez Gene ID
2549
Gene Name
GRB2 associated binding protein 1
Gene Aliases
-
Chromosome Location
Chr.4: 143336830 - 143474565 on GRCh38
UniGene ID
Hs.618456
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_002039.3 NP_002030.2 2
NM_207123.2 NP_997006.1 2
XM_006714167.3 XP_006714230.1 2
XM_006714168.3 XP_006714231.1 2
XM_017007966.1 XP_016863455.1 3
XM_017007967.1 XP_016863456.1 2
XM_017007968.1 XP_016863457.1 3
XM_017007969.1 XP_016863458.1 2
XM_017007970.1 XP_016863459.1 2
XM_017007971.1 XP_016863460.1 2