gRNA Design Details

Target DNA Sequence
CGAACGGGTAGAAGTCGGCA
PAM Sequence
AGG
Target locus
Chr.2: 240998918 - 240998940 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 94.82/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
SNED1
Entrez Gene ID
25992
Gene Name
sushi, nidogen and EGF like domains 1
Gene Aliases
SST3, Snep
Chromosome Location
Chr.2: 240997409 - 241095568 on GRCh38
UniGene ID
Hs.471834
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001080437.1 NP_001073906.1 1
XM_017003774.1 XP_016859263.1 1