gRNA Design Details

Target DNA Sequence
GAGAAATCTATACCATCCAG
PAM Sequence
GGG
Target locus
Chr.7: 12393636 - 12393658 on GRCh38
Strand
Forward
Application
Gene Knock-Out
Score: 90.18/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
VWDE
Entrez Gene ID
221806
Gene Name
von Willebrand factor D and EGF domains
Gene Aliases
-
Chromosome Location
Chr.7: 12330883 - 12404226 on GRCh38
UniGene ID
Hs.669526
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001135924.1 NP_001129396.1 2
XM_005249641.1 XP_005249698.1 2
XM_011515174.1 XP_011513476.1 2
XM_011515175.1 XP_011513477.1 2
XM_017011828.1 XP_016867317.1 2
XM_017011830.1 XP_016867319.1 2