gRNA Design Details

Target DNA Sequence
AGACCAGGTGCAGCACTTCA
PAM Sequence
AGG
Target locus
Chr.17: 19024355 - 19024377 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 68.67/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
GRAP
Entrez Gene ID
10750
Gene Name
GRB2-related adaptor protein
Gene Aliases
-
Chromosome Location
Chr.17: 19020656 - 19051352 on GRCh38
UniGene ID
Hs.567416
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_006613.3 NP_006604.1 4
XM_017024029.1 XP_016879518.1 5