gRNA Design Details

Target DNA Sequence
AGTCGCTGTTGGGTCTCATG
PAM Sequence
GGG
Target locus
Chr.5: 38451396 - 38451418 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 91.09/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
EGFLAM
Entrez Gene ID
133584
Gene Name
EGF like, fibronectin type III and laminin G domains
Gene Aliases
AGRINL, AGRNL, PIKA
Chromosome Location
Chr.5: 38258409 - 38465480 on GRCh38
UniGene ID
Hs.20103
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001205301.1 NP_001192230.1 20
NM_152403.3 NP_689616.2 19
NM_182798.2 NP_877950.1 14
NM_182801.2 NP_877953.1 3