gRNA Design Details
- Target DNA Sequence
- ATGCGGCAGCTTAACATCAA
- PAM Sequence
- TGG
- Target locus
-
Chr.5: 38462975 - 38462997 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
89.52/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- EGFLAM
- Entrez Gene ID
-
133584
- Gene Name
- EGF like, fibronectin type III and laminin G domains
- Gene Aliases
-
AGRINL, AGRNL, PIKA
- Chromosome Location
-
Chr.5: 38258409 - 38465480 on GRCh38
- UniGene ID
- Hs.20103
- Species
-
Human