Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000662.7 | NP_000653.3 | 3 | 307 | |
NM_001160170.3 | NP_001153642.1 | 5 | 841 | ||
NM_001160171.3 | NP_001153643.1 | 4 | 691 | ||
NM_001160172.3 | NP_001153644.1 | 5 | 612 | ||
NM_001160173.3 | NP_001153645.1 | 4 | 457 | ||
NM_001160174.2 | NP_001153646.1 | 1 | 524 | ||
NM_001160175.3 | NP_001153647.1 | 5 | 575 | ||
NM_001160176.3 | NP_001153648.1 | 4 | 425 | ||
NM_001160179.2 | NP_001153651.1 | 5 | 511 | ||
NM_001291962.1 | NP_001278891.1 | 6 | 629 | ||
XM_006716410.3 | XP_006716473.1 | 3 | 884 | ||
XM_011544687.1 | XP_011542989.1 | 6 | 970 | ||
XM_011544688.1 | XP_011542990.1 | 5 | 820 | ||
XM_011544689.2 | XP_011542991.1 | 2 | 669 | ||
XM_017013947.1 | XP_016869436.1 | 8 | 1206 | ||
GenBank mRNA | AJ581135.1 | - | 3 | 271 | |
AJ581136.1 | - | 3 | 287 | ||
AJ581137.1 | - | 4 | 442 | ||
AJ581138.1 | - | 4 | 672 | ||
AJ581139.1 | - | 4 | 405 | ||
AJ581140.1 | - | 2 | 194 | ||
AJ581141.1 | - | 1 | 524 | ||
AK290912.1 | - | 3 | 782 | ||
AV754344.1 | - | 3 | 304 | ||
BC013732.1 | - | 4 | 664 | ||
BC047666.1 | - | 3 | 286 | 6059458 | |
BG655073.1 | - | 3 | 334 | ||
BM924372.1 | - | 5 | 586 | ||
BX647521.1 | - | 4 | 400 | ||
CD702544.1 | - | 5 | 523 | ||
D90041.1 | - | 1 | 290 |