Gene Transcripts
- Gene Symbol
- KIAA1033
- Entrez Gene ID
- 23325
- Gene Name
- KIAA1033
- Gene Aliases
- MRT43, SWIP
- Chromosome Location
- Chr. 12: 105107714 - 105169134 on Build GRCh38
- UniGene ID
- Hs.12144
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001293640.1 |
NP_001280569.1
|
31, 32 |
3439 |
|
NM_015275.2 |
NP_056090.1
|
31, 32 |
3436 |
|
XM_011538073.2 |
XP_011536375.1
|
30, 31 |
3301 |
|
XM_011538074.2 |
XP_011536376.1
|
31, 32 |
3402 |
|
GenBank mRNA |
AB028956.1 |
- |
31, 32 |
3418 |
|
AL137753.1 |
- |
28, 29 |
3060 |
|
BC031358.1 |
- |
2, 3 |
278 |
|
BC040936.1 |
- |
8, 9 |
950 |
|
BC104992.1 |
- |
31, 32 |
3378 |
8143995 |
BC104994.1 |
- |
31, 32 |
3378 |
8143997 |
BC143373.1 |
- |
31, 32 |
3381 |
9051878 |
View Full Product Details