Gene Transcripts
- Gene Symbol
- TAS1R2
- Entrez Gene ID
- 80834
- Gene Name
- taste 1 receptor member 2
- Gene Aliases
- GPR71, T1R2, TR2
- Chromosome Location
- Chr. 1: 18839599 - 18859661 on Build GRCh38
- UniGene ID
- Hs.553548
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_152232.2 |
NP_689418.2
|
6 |
2041 |
|
View Full Product Details