Gene Transcripts

Gene Symbol
ZGRF1
Entrez Gene ID
55345
Gene Name
zinc finger GRF-type containing 1
Gene Aliases
C4orf21
Chromosome Location
Chr. 4: 112539333 - 112636995 on Build GRCh38
UniGene ID
Hs.380346
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018392.4 NP_060862.3 23 5570
XM_005263115.3 XP_005263172.1 23 5625
XM_011532091.2 XP_011530393.1 22 5519
XM_011532092.2 XP_011530394.1 22 5450
XM_011532093.2 XP_011530395.1 22 5396
XM_011532094.2 XP_011530396.1 21 5312
XM_011532097.2 XP_011530399.1 16 2672
XM_011532098.2 XP_011530400.1 14 2346
XM_011532099.2 XP_011530401.1 13 2305
XM_017008369.1 XP_016863858.1 22 5565
XM_017008370.1 XP_016863859.1 21 5345
XM_017008371.1 XP_016863860.1 20 5225
XM_017008372.1 XP_016863861.1 20 4604
XM_017008373.1 XP_016863862.1 19 4541
GenBank mRNA AK090556.1 - 5 1205
AK126054.1 - 12 1954

View Full Product Details