Gene Transcripts
- Gene Symbol
- LINC00152
- Entrez Gene ID
- 112597
- Gene Name
- long intergenic non-protein coding RNA 152
- Gene Aliases
- C2orf59, NCRNA00152
- Chromosome Location
- Chr. 2: 87455455 - 87521518 on Build GRCh38
- UniGene ID
- Hs.652166
- Species
- Homo sapiens
- Gene Symbol
- MIR4435-2HG
- Entrez Gene ID
- 541471
- Gene Name
- MIR4435-2 host gene
- Gene Aliases
- AGD2, AK001796, LINC00978, MIR4435-1HG
- Chromosome Location
- Chr. 2: 111195866 - 111495161 on Build GRCh38
- UniGene ID
- Hs.560805
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024373.2 |
|
2 |
501 |
|
GenBank mRNA |
BC014776.1 |
- |
2 |
428 |
|
BC070201.1 |
- |
2 |
388 |
|
View Full Product Details