Gene Transcripts

Gene Symbol
PPM1K
Entrez Gene ID
152926
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1K
Gene Aliases
BDP, MSUDMV, PP2Ckappa, PP2Cm, PTMP, UG0882E07
Chromosome Location
Chr. 4: 88257609 - 88284831 on Build GRCh38
UniGene ID
Hs.43744
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152542.4 NP_689755.3 2 838
XM_005262775.3 XP_005262832.1 2 636
XM_006714111.3 XP_006714174.1 2 899
XM_017007803.1 XP_016863292.1 2 486
XM_017007804.1 XP_016863293.1 2 833
XM_017007805.1 XP_016863294.1 2 833
GenBank mRNA AF351614.1 - 2 569
AK054678.1 - 2 566
AK314417.1 - 2 563
AL834167.1 - 2 776
AL834271.1 - 2 557
AX766363.1 - 1 353
AY157615.1 - 2 566
AY435431.1 - 2 743
AY994097.1 - 2 574
BC020850.1 - 2 568
BC037552.1 - 2 602 5227317
BC041350.1 - 1 1474

View Full Product Details