Gene Transcripts
- Gene Symbol
- WDFY3
- Entrez Gene ID
- 23001
- Gene Name
- WD repeat and FYVE domain containing 3
- Gene Aliases
- ALFY, ZFYVE25
- Chromosome Location
- Chr. 4: 84669537 - 84966391 on Build GRCh38
- UniGene ID
- Hs.480116
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_014991.4 |
NP_055806.2
|
29, 30 |
5170 |
|
XM_005262858.4 |
XP_005262915.1
|
31, 32 |
5211 |
|
XM_011531757.2 |
XP_011530059.1
|
30, 31 |
5122 |
|
XM_011531759.2 |
XP_011530061.1
|
29, 30 |
5084 |
|
XM_011531760.2 |
XP_011530062.1
|
29, 30 |
5022 |
|
XM_011531761.2 |
XP_011530063.1
|
30, 31 |
5122 |
|
XM_011531762.2 |
XP_011530064.1
|
30, 31 |
5122 |
|
XM_011531763.2 |
XP_011530065.1
|
28, 29 |
4968 |
|
XM_011531764.2 |
XP_011530066.1
|
29, 30 |
5038 |
|
XM_011531765.2 |
XP_011530067.1
|
27, 28 |
4824 |
|
XM_011531766.2 |
XP_011530068.1
|
30, 31 |
5122 |
|
XM_011531767.2 |
XP_011530069.1
|
30, 31 |
5122 |
|
XM_017007906.1 |
XP_016863395.1
|
32, 33 |
5339 |
|
XM_017007907.1 |
XP_016863396.1
|
32, 33 |
5336 |
|
XM_017007908.1 |
XP_016863397.1
|
31, 32 |
5268 |
|
XM_017007909.1 |
XP_016863398.1
|
17, 18 |
2964 |
|
GenBank mRNA |
AF538685.1 |
- |
28, 29 |
5062 |
|
View Full Product Details