Gene Transcripts

Gene Symbol
CACNB2
Entrez Gene ID
783
Gene Name
calcium voltage-gated channel auxiliary subunit beta 2
Gene Aliases
CACNLB2, CAVB2, MYSB
Chromosome Location
Chr. 10: 18140560 - 18545881 on Build GRCh38
UniGene ID
Hs.59093
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000724.3 NP_000715.2 4 929
NM_001167945.1 NP_001161417.1 5 762
NM_201570.2 NP_963864.1 4 783
NM_201571.3 NP_963865.2 5 762
NM_201572.3 NP_963866.2 5 762
NM_201590.2 NP_963884.2 4 559
NM_201593.2 NP_963887.2 5 539
NM_201596.2 NP_963890.2 5 539
NM_201597.2 NP_963891.1 5 539
XM_005252588.3 XP_005252645.1 4 787
XM_006717502.3 XP_006717565.1 4 592
XM_011519659.2 XP_011517961.1 4 559
XM_011519660.2 XP_011517962.1 4 1144
XM_017016625.1 XP_016872114.1 4 699
GenBank mRNA AB208917.1 - 4 815
AF137376.1 - 5 477
AF137377.1 - 4 320
AF285239.2 - 4 317
AF423189.1 - 4 314
AF423190.1 - 5 395
AF423191.1 - 5 479
AF423192.1 - 4 335
AF465485.1 - 4 323
AK293837.1 - 5 694
AK293937.1 - 4 344
AK295113.1 - 4 680
AK296471.1 - 5 446
AK299873.1 - 4 685
AK307526.1 - 5 762
AK307800.1 - 4 505
AK310943.1 - 4 559
AL119895.1 - 5 427
AL600557.1 - 4 485
AY393858.1 - 5 395
AY393859.1 - 4 317
AY393860.1 - 5 479
AY393861.1 - 5 395
AY393862.1 - 4 317
AY393863.1 - 4 317
BC136409.1 - 5 502 9020397
BM312516.1 - 2 46
S60415.1 - 5 539
U95019.1 - 4 815

View Full Product Details