Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 15483114-15483537 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CELA2B
Entrez Gene ID
51032
Gene Name
chymotrypsin like elastase family member 2B
Gene Aliases
ELA2B
UniGene
Hs.631871
Gene Chromosome Location
Chr.1: 15476101-15491400 on Build GRCh38
Transcripts
NM_015849.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
897668 c.469G>A p.V157I Chr.1: 15483376-15483376 on Build GRCh38
4023630 c.360C>T p.N120N Chr.1: 15483267-15483267 on Build GRCh38
1295276 c.399C>T p.L133L Chr.1: 15483306-15483306 on Build GRCh38
6058445 c.389C>A p.P130H Chr.1: 15483296-15483296 on Build GRCh38
1721681 c.390C>T p.P130P Chr.1: 15483297-15483297 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs12129367 Chr.1: 15483211-15483211 on Build GRCh38
rs752727312 Chr.1: 15483274-15483274 on Build GRCh38
rs143869575 Chr.1: 15483168-15483168 on Build GRCh38
rs752676862 Chr.1: 15483343-15483343 on Build GRCh38
rs754897394 Chr.1: 15483268-15483268 on Build GRCh38
rs1029072615 Chr.1: 15483206-15483206 on Build GRCh38
rs533711715 Chr.1: 15483244-15483244 on Build GRCh38
rs575698771 Chr.1: 15483508-15483508 on Build GRCh38
rs745586197 Chr.1: 15483380-15483380 on Build GRCh38
rs756927007 Chr.1: 15483375-15483375 on Build GRCh38
rs745398470 Chr.1: 15483296-15483296 on Build GRCh38
rs748716056 Chr.1: 15483305-15483305 on Build GRCh38
rs1015181902 Chr.1: 15483167-15483168 on Build GRCh38
rs762271458 Chr.1: 15483462-15483462 on Build GRCh38
rs1004777605 Chr.1: 15483159-15483159 on Build GRCh38
rs535516211 Chr.1: 15483470-15483470 on Build GRCh38
rs747407535 Chr.1: 15483422-15483422 on Build GRCh38
rs940334551 Chr.1: 15483408-15483408 on Build GRCh38
rs201996525 Chr.1: 15483233-15483233 on Build GRCh38
rs746589408 Chr.1: 15483286-15483286 on Build GRCh38
rs780674401 Chr.1: 15483376-15483376 on Build GRCh38
rs12726692 Chr.1: 15483141-15483141 on Build GRCh38
rs575781422 Chr.1: 15483232-15483232 on Build GRCh38
rs772582382 Chr.1: 15483400-15483400 on Build GRCh38
rs369813912 Chr.1: 15483347-15483347 on Build GRCh38
rs759869086 Chr.1: 15483424-15483424 on Build GRCh38
rs149546145 Chr.1: 15483157-15483157 on Build GRCh38
rs564766934 Chr.1: 15483236-15483236 on Build GRCh38
rs774701588 Chr.1: 15483304-15483304 on Build GRCh38
rs139801516 Chr.1: 15483326-15483326 on Build GRCh38
rs781195609 Chr.1: 15483412-15483420 on Build GRCh38
rs763455582 Chr.1: 15483240-15483240 on Build GRCh38
rs774735622 Chr.1: 15483245-15483245 on Build GRCh38
rs928863614 Chr.1: 15483308-15483308 on Build GRCh38
rs751418118 Chr.1: 15483372-15483372 on Build GRCh38
rs544510712 Chr.1: 15483235-15483235 on Build GRCh38
rs777778103 Chr.1: 15483279-15483279 on Build GRCh38
rs184117904 Chr.1: 15483435-15483435 on Build GRCh38
rs773518091 Chr.1: 15483420-15483420 on Build GRCh38
rs199989062 Chr.1: 15483267-15483267 on Build GRCh38
rs756982665 Chr.1: 15483451-15483451 on Build GRCh38
rs1015880559 Chr.1: 15483138-15483138 on Build GRCh38
rs778683649 Chr.1: 15483269-15483269 on Build GRCh38
rs376677025 Chr.1: 15483344-15483344 on Build GRCh38
rs938903556 Chr.1: 15483309-15483309 on Build GRCh38
rs772031053 Chr.1: 15483216-15483216 on Build GRCh38
rs189924451 Chr.1: 15483461-15483461 on Build GRCh38
rs373971727 Chr.1: 15483423-15483423 on Build GRCh38
rs10927792 Chr.1: 15483381-15483381 on Build GRCh38
rs542332918 Chr.1: 15483175-15483175 on Build GRCh38
rs552957433 Chr.1: 15483167-15483167 on Build GRCh38
rs773571189 Chr.1: 15483310-15483310 on Build GRCh38
rs779686295 Chr.1: 15483388-15483388 on Build GRCh38
rs766534078 Chr.1: 15483315-15483315 on Build GRCh38
rs748587640 Chr.1: 15483393-15483393 on Build GRCh38
rs745659815 Chr.1: 15483430-15483430 on Build GRCh38
rs72866915 Chr.1: 15483246-15483246 on Build GRCh38
rs758253187 Chr.1: 15483278-15483278 on Build GRCh38
rs769388813 Chr.1: 15483298-15483298 on Build GRCh38
rs201296279 Chr.1: 15483290-15483290 on Build GRCh38
rs577560545 Chr.1: 15483146-15483146 on Build GRCh38
rs544785659 Chr.1: 15483511-15483511 on Build GRCh38
rs767922169 Chr.1: 15483251-15483251 on Build GRCh38
rs200820219 Chr.1: 15483306-15483306 on Build GRCh38
rs977731534 Chr.1: 15483272-15483272 on Build GRCh38
rs200410579 Chr.1: 15483396-15483396 on Build GRCh38
rs533837250 Chr.1: 15483432-15483432 on Build GRCh38
rs746648292 Chr.1: 15483224-15483224 on Build GRCh38
rs769415604 Chr.1: 15483451-15483452 on Build GRCh38
rs764155845 Chr.1: 15483433-15483433 on Build GRCh38
rs571165671 Chr.1: 15483429-15483429 on Build GRCh38
rs770960163 Chr.1: 15483323-15483324 on Build GRCh38
rs757905911 Chr.1: 15483139-15483139 on Build GRCh38
rs760954295 Chr.1: 15483257-15483257 on Build GRCh38
rs538038132 Chr.1: 15483349-15483349 on Build GRCh38
rs765414243 Chr.1: 15483342-15483342 on Build GRCh38
rs775720693 Chr.1: 15483431-15483431 on Build GRCh38
rs770716972 Chr.1: 15483289-15483289 on Build GRCh38
rs371785024 Chr.1: 15483252-15483252 on Build GRCh38
rs181422030 Chr.1: 15483499-15483499 on Build GRCh38
rs776733977 Chr.1: 15483318-15483318 on Build GRCh38

View Full Product Details