Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 49884791-49885292 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALG12
Entrez Gene ID
79087
Gene Name
ALG12, alpha-1,6-mannosyltransferase
Gene Aliases
CDG1G, ECM39, PP14673, hALG12
UniGene
Hs.526711
Gene Chromosome Location
Chr.22: 49860163-49918469 on Build GRCh38
Transcripts
XM_017028936.1 XM_017028937.1 NM_014838.2

Gene Symbol
ZBED4
Entrez Gene ID
9889
Gene Name
zinc finger BED-type containing 4
Gene Aliases
-
UniGene
Hs.475208
Gene Chromosome Location
Chr.22: 49853849-49890078 on Build GRCh38
Transcripts
XM_017028936.1 XM_017028937.1 NM_014838.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7003090 c.1550G>A p.G517D Chr.22: 49885212-49885212 on Build GRCh38
4843008 c.1154C>T p.S385L Chr.22: 49884816-49884816 on Build GRCh38
298410 c.1319T>G p.L440W Chr.22: 49884981-49884981 on Build GRCh38
4156406 c.1258A>G p.I420V Chr.22: 49884920-49884920 on Build GRCh38
1416943 c.1361T>C p.M454T Chr.22: 49885023-49885023 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755197118 Chr.22: 49884818-49884818 on Build GRCh38
rs780126311 Chr.22: 49884816-49884816 on Build GRCh38
rs976694901 Chr.22: 49885101-49885101 on Build GRCh38
rs765485064 Chr.22: 49885212-49885212 on Build GRCh38
rs369830557 Chr.22: 49885131-49885131 on Build GRCh38
rs910800 Chr.22: 49884994-49884994 on Build GRCh38
rs911397457 Chr.22: 49884996-49884996 on Build GRCh38
rs910799 Chr.22: 49884920-49884920 on Build GRCh38
rs772724161 Chr.22: 49885075-49885075 on Build GRCh38
rs757888454 Chr.22: 49885161-49885182 on Build GRCh38
rs764054268 Chr.22: 49885207-49885207 on Build GRCh38
rs957549002 Chr.22: 49885004-49885004 on Build GRCh38
rs749701905 Chr.22: 49885079-49885079 on Build GRCh38
rs922581459 Chr.22: 49885041-49885041 on Build GRCh38
rs201032978 Chr.22: 49884862-49884862 on Build GRCh38
rs747541628 Chr.22: 49885050-49885050 on Build GRCh38
rs1049898818 Chr.22: 49884924-49884924 on Build GRCh38
rs776412890 Chr.22: 49884892-49884892 on Build GRCh38
rs773627861 Chr.22: 49884943-49884943 on Build GRCh38
rs769556083 Chr.22: 49885053-49885053 on Build GRCh38
rs146505525 Chr.22: 49884979-49884979 on Build GRCh38
rs368514449 Chr.22: 49885257-49885257 on Build GRCh38
rs778790812 Chr.22: 49884928-49884928 on Build GRCh38
rs756347823 Chr.22: 49885189-49885189 on Build GRCh38
rs766928121 Chr.22: 49884968-49884968 on Build GRCh38
rs761545349 Chr.22: 49884937-49884937 on Build GRCh38
rs201109219 Chr.22: 49884897-49884897 on Build GRCh38
rs781388058 Chr.22: 49884825-49884825 on Build GRCh38
rs776010776 Chr.22: 49885022-49885022 on Build GRCh38
rs141092774 Chr.22: 49885190-49885190 on Build GRCh38
rs146411541 Chr.22: 49884936-49884936 on Build GRCh38
rs746916270 Chr.22: 49885019-49885019 on Build GRCh38
rs781440994 Chr.22: 49884914-49884914 on Build GRCh38
rs373015001 Chr.22: 49884906-49884906 on Build GRCh38
rs767616881 Chr.22: 49884974-49884974 on Build GRCh38
rs752544877 Chr.22: 49885228-49885228 on Build GRCh38
rs775155655 Chr.22: 49884869-49884869 on Build GRCh38
rs747750398 Chr.22: 49885151-49885151 on Build GRCh38
rs780762652 Chr.22: 49885039-49885039 on Build GRCh38
rs745593833 Chr.22: 49884931-49884931 on Build GRCh38
rs202182491 Chr.22: 49884913-49884913 on Build GRCh38
rs773947931 Chr.22: 49885220-49885220 on Build GRCh38
rs1024073527 Chr.22: 49884964-49884964 on Build GRCh38
rs888614792 Chr.22: 49885129-49885129 on Build GRCh38
rs116893184 Chr.22: 49885162-49885162 on Build GRCh38
rs778757442 Chr.22: 49884841-49884841 on Build GRCh38
rs61731525 Chr.22: 49885231-49885231 on Build GRCh38
rs767538197 Chr.22: 49885222-49885222 on Build GRCh38
rs913929434 Chr.22: 49885060-49885060 on Build GRCh38
rs945368013 Chr.22: 49885085-49885085 on Build GRCh38
rs1054202373 Chr.22: 49884874-49884874 on Build GRCh38
rs771253063 Chr.22: 49885083-49885083 on Build GRCh38
rs777170849 Chr.22: 49885233-49885233 on Build GRCh38
rs375447880 Chr.22: 49884967-49884967 on Build GRCh38
rs759390898 Chr.22: 49885218-49885218 on Build GRCh38
rs748969694 Chr.22: 49885169-49885169 on Build GRCh38
rs200066649 Chr.22: 49884831-49884831 on Build GRCh38
rs143395671 Chr.22: 49884883-49884883 on Build GRCh38
rs756909589 Chr.22: 49885254-49885254 on Build GRCh38
rs767856195 Chr.22: 49885095-49885095 on Build GRCh38
rs750301949 Chr.22: 49884993-49884993 on Build GRCh38
rs750699650 Chr.22: 49884901-49884901 on Build GRCh38
rs751704901 Chr.22: 49884912-49884912 on Build GRCh38
rs751578085 Chr.22: 49884817-49884817 on Build GRCh38
rs151212072 Chr.22: 49884893-49884893 on Build GRCh38
rs760058831 Chr.22: 49885088-49885088 on Build GRCh38
rs771856285 Chr.22: 49884868-49884868 on Build GRCh38
rs775850873 Chr.22: 49885181-49885181 on Build GRCh38
rs184545959 Chr.22: 49884898-49884898 on Build GRCh38
rs752933692 Chr.22: 49884919-49884919 on Build GRCh38
rs137870205 Chr.22: 49885234-49885234 on Build GRCh38
rs201983662 Chr.22: 49885107-49885107 on Build GRCh38
rs143765058 Chr.22: 49885126-49885126 on Build GRCh38
rs544082416 Chr.22: 49884983-49884983 on Build GRCh38
rs770736620 Chr.22: 49884835-49884835 on Build GRCh38
rs374406255 Chr.22: 49885163-49885163 on Build GRCh38
rs1020525364 Chr.22: 49884981-49884981 on Build GRCh38
rs746167202 Chr.22: 49885173-49885173 on Build GRCh38
rs149970137 Chr.22: 49884933-49884933 on Build GRCh38
rs763525210 Chr.22: 49884902-49884902 on Build GRCh38
rs770523019 Chr.22: 49885170-49885170 on Build GRCh38
rs200609130 Chr.22: 49885097-49885097 on Build GRCh38
rs759652014 Chr.22: 49884969-49884969 on Build GRCh38
rs753989665 Chr.22: 49885114-49885114 on Build GRCh38
rs202162671 Chr.22: 49885102-49885102 on Build GRCh38
rs969381349 Chr.22: 49884989-49884989 on Build GRCh38
rs1009151518 Chr.22: 49885194-49885194 on Build GRCh38
rs778534074 Chr.22: 49885255-49885255 on Build GRCh38
rs61731526 Chr.22: 49884940-49884940 on Build GRCh38
rs760200733 Chr.22: 49884876-49884876 on Build GRCh38
rs766803700 Chr.22: 49884904-49884904 on Build GRCh38
rs760669604 Chr.22: 49885103-49885103 on Build GRCh38
rs370021114 Chr.22: 49885118-49885118 on Build GRCh38
rs752269506 Chr.22: 49885103-49885103 on Build GRCh38
rs377369650 Chr.22: 49884978-49884978 on Build GRCh38
rs141708563 Chr.22: 49884997-49884997 on Build GRCh38
rs774711027 Chr.22: 49885171-49885171 on Build GRCh38
rs751464773 Chr.22: 49885214-49885214 on Build GRCh38
rs772704581 Chr.22: 49885176-49885176 on Build GRCh38
rs776815347 Chr.22: 49885211-49885211 on Build GRCh38
rs550777671 Chr.22: 49885119-49885119 on Build GRCh38
rs770046152 Chr.22: 49885021-49885021 on Build GRCh38
rs780984672 Chr.22: 49885147-49885147 on Build GRCh38
rs758312324 Chr.22: 49885117-49885117 on Build GRCh38
rs370628347 Chr.22: 49884934-49884934 on Build GRCh38
rs774659625 Chr.22: 49885087-49885087 on Build GRCh38
rs764157053 Chr.22: 49884985-49884985 on Build GRCh38

View Full Product Details