Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 49885179-49885689 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALG12
Entrez Gene ID
79087
Gene Name
ALG12, alpha-1,6-mannosyltransferase
Gene Aliases
CDG1G, ECM39, PP14673, hALG12
UniGene
Hs.526711
Gene Chromosome Location
Chr.22: 49860163-49918469 on Build GRCh38
Transcripts
XM_017028936.1 XM_017028937.1 NM_014838.2

Gene Symbol
ZBED4
Entrez Gene ID
9889
Gene Name
zinc finger BED-type containing 4
Gene Aliases
-
UniGene
Hs.475208
Gene Chromosome Location
Chr.22: 49853849-49890078 on Build GRCh38
Transcripts
XM_017028936.1 XM_017028937.1 NM_014838.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
479098 c.1766G>C p.G589A Chr.22: 49885428-49885428 on Build GRCh38
7003090 c.1550G>A p.G517D Chr.22: 49885212-49885212 on Build GRCh38
267884 c.1714G>A p.A572T Chr.22: 49885376-49885376 on Build GRCh38
5728609 c.2017T>C p.Y673H Chr.22: 49885679-49885679 on Build GRCh38
7114518 c.1798C>G p.L600V Chr.22: 49885460-49885460 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs571708422 Chr.22: 49885408-49885408 on Build GRCh38
rs759274549 Chr.22: 49885609-49885609 on Build GRCh38
rs566333083 Chr.22: 49885496-49885496 on Build GRCh38
rs746830307 Chr.22: 49885523-49885523 on Build GRCh38
rs150596388 Chr.22: 49885518-49885518 on Build GRCh38
rs775153174 Chr.22: 49885416-49885416 on Build GRCh38
rs765485064 Chr.22: 49885212-49885212 on Build GRCh38
rs145958586 Chr.22: 49885300-49885300 on Build GRCh38
rs201721474 Chr.22: 49885308-49885308 on Build GRCh38
rs112605283 Chr.22: 49885291-49885291 on Build GRCh38
rs764054268 Chr.22: 49885207-49885207 on Build GRCh38
rs766101470 Chr.22: 49885375-49885375 on Build GRCh38
rs142674632 Chr.22: 49885438-49885438 on Build GRCh38
rs371316523 Chr.22: 49885574-49885574 on Build GRCh38
rs749387965 Chr.22: 49885560-49885560 on Build GRCh38
rs577430389 Chr.22: 49885552-49885552 on Build GRCh38
rs368514449 Chr.22: 49885257-49885257 on Build GRCh38
rs141697372 Chr.22: 49885365-49885365 on Build GRCh38
rs199728678 Chr.22: 49885544-49885544 on Build GRCh38
rs773175941 Chr.22: 49885581-49885581 on Build GRCh38
rs769106960 Chr.22: 49885394-49885394 on Build GRCh38
rs200511541 Chr.22: 49885323-49885323 on Build GRCh38
rs144634675 Chr.22: 49885536-49885536 on Build GRCh38
rs757339917 Chr.22: 49885564-49885564 on Build GRCh38
rs761765426 Chr.22: 49885529-49885529 on Build GRCh38
rs752544877 Chr.22: 49885228-49885228 on Build GRCh38
rs756748204 Chr.22: 49885376-49885376 on Build GRCh38
rs202191291 Chr.22: 49885299-49885299 on Build GRCh38
rs144547756 Chr.22: 49885413-49885413 on Build GRCh38
rs748284131 Chr.22: 49885396-49885396 on Build GRCh38
rs543682137 Chr.22: 49885474-49885474 on Build GRCh38
rs751923383 Chr.22: 49885463-49885463 on Build GRCh38
rs773947931 Chr.22: 49885220-49885220 on Build GRCh38
rs148466799 Chr.22: 49885437-49885437 on Build GRCh38
rs371556038 Chr.22: 49885479-49885479 on Build GRCh38
rs200415861 Chr.22: 49885395-49885395 on Build GRCh38
rs746136211 Chr.22: 49885568-49885568 on Build GRCh38
rs764079284 Chr.22: 49885423-49885423 on Build GRCh38
rs747504763 Chr.22: 49885282-49885282 on Build GRCh38
rs374263929 Chr.22: 49885435-49885435 on Build GRCh38
rs79032526 Chr.22: 49885489-49885489 on Build GRCh38
rs772660581 Chr.22: 49885537-49885537 on Build GRCh38
rs61731525 Chr.22: 49885231-49885231 on Build GRCh38
rs767538197 Chr.22: 49885222-49885222 on Build GRCh38
rs773684685 Chr.22: 49885596-49885596 on Build GRCh38
rs776530063 Chr.22: 49885425-49885425 on Build GRCh38
rs767211068 Chr.22: 49885663-49885663 on Build GRCh38
rs776372775 Chr.22: 49885525-49885525 on Build GRCh38
rs770503857 Chr.22: 49885594-49885594 on Build GRCh38
rs368061670 Chr.22: 49885551-49885551 on Build GRCh38
rs760544641 Chr.22: 49885367-49885367 on Build GRCh38
rs777170849 Chr.22: 49885233-49885233 on Build GRCh38
rs759390898 Chr.22: 49885218-49885218 on Build GRCh38
rs778589268 Chr.22: 49885378-49885378 on Build GRCh38
rs760515369 Chr.22: 49885418-49885418 on Build GRCh38
rs756909589 Chr.22: 49885254-49885254 on Build GRCh38
rs557091777 Chr.22: 49885461-49885461 on Build GRCh38
rs778012406 Chr.22: 49885492-49885492 on Build GRCh38
rs922716056 Chr.22: 49885320-49885320 on Build GRCh38
rs926982020 Chr.22: 49885587-49885587 on Build GRCh38
rs772736054 Chr.22: 49885285-49885285 on Build GRCh38
rs368462627 Chr.22: 49885441-49885441 on Build GRCh38
rs763952816 Chr.22: 49885373-49885373 on Build GRCh38
rs772304085 Chr.22: 49885271-49885271 on Build GRCh38
rs137870205 Chr.22: 49885234-49885234 on Build GRCh38
rs199668546 Chr.22: 49885549-49885549 on Build GRCh38
rs571481002 Chr.22: 49885316-49885316 on Build GRCh38
rs778340747 Chr.22: 49885554-49885554 on Build GRCh38
rs780308372 Chr.22: 49885391-49885391 on Build GRCh38
rs199867332 Chr.22: 49885305-49885305 on Build GRCh38
rs997382257 Chr.22: 49885513-49885513 on Build GRCh38
rs937036902 Chr.22: 49885610-49885610 on Build GRCh38
rs138599627 Chr.22: 49885345-49885345 on Build GRCh38
rs200372064 Chr.22: 49885545-49885545 on Build GRCh38
rs965944100 Chr.22: 49885495-49885495 on Build GRCh38
rs530191361 Chr.22: 49885272-49885272 on Build GRCh38
rs779045858 Chr.22: 49885567-49885567 on Build GRCh38
rs765993642 Chr.22: 49885542-49885542 on Build GRCh38
rs750048993 Chr.22: 49885386-49885386 on Build GRCh38
rs747328403 Chr.22: 49885393-49885393 on Build GRCh38
rs778534074 Chr.22: 49885255-49885255 on Build GRCh38
rs145536914 Chr.22: 49885387-49885387 on Build GRCh38
rs976724304 Chr.22: 49885322-49885322 on Build GRCh38
rs756693965 Chr.22: 49885553-49885553 on Build GRCh38
rs1020038365 Chr.22: 49885493-49885493 on Build GRCh38
rs756638938 Chr.22: 49885488-49885488 on Build GRCh38
rs749323294 Chr.22: 49885512-49885512 on Build GRCh38
rs753632021 Chr.22: 49885519-49885519 on Build GRCh38
rs149686381 Chr.22: 49885288-49885288 on Build GRCh38
rs751464773 Chr.22: 49885214-49885214 on Build GRCh38
rs776815347 Chr.22: 49885211-49885211 on Build GRCh38
rs772237818 Chr.22: 49885572-49885572 on Build GRCh38
rs773385687 Chr.22: 49885410-49885410 on Build GRCh38
rs762408365 Chr.22: 49885586-49885586 on Build GRCh38
rs201734469 Chr.22: 49885412-49885412 on Build GRCh38
rs200730742 Chr.22: 49885573-49885573 on Build GRCh38
rs768410847 Chr.22: 49885524-49885524 on Build GRCh38
rs781774175 Chr.22: 49885478-49885478 on Build GRCh38
rs766047605 Chr.22: 49885447-49885447 on Build GRCh38

View Full Product Details