Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 49884579-49884852 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALG12
Entrez Gene ID
79087
Gene Name
ALG12, alpha-1,6-mannosyltransferase
Gene Aliases
CDG1G, ECM39, PP14673, hALG12
UniGene
Hs.526711
Gene Chromosome Location
Chr.22: 49860163-49918469 on Build GRCh38
Transcripts
XM_017028936.1 XM_017028937.1 NM_014838.2

Gene Symbol
ZBED4
Entrez Gene ID
9889
Gene Name
zinc finger BED-type containing 4
Gene Aliases
-
UniGene
Hs.475208
Gene Chromosome Location
Chr.22: 49853849-49890078 on Build GRCh38
Transcripts
XM_017028936.1 XM_017028937.1 NM_014838.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6890499 c.1118C>T p.S373F Chr.22: 49884780-49884780 on Build GRCh38
5458955 c.1037C>T p.T346M Chr.22: 49884699-49884699 on Build GRCh38
4843008 c.1154C>T p.S385L Chr.22: 49884816-49884816 on Build GRCh38
4582495 c.1047G>A p.P349P Chr.22: 49884709-49884709 on Build GRCh38
1641582 c.949A>G p.K317E Chr.22: 49884611-49884611 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369020472 Chr.22: 49884691-49884691 on Build GRCh38
rs753177858 Chr.22: 49884671-49884671 on Build GRCh38
rs758495510 Chr.22: 49884757-49884757 on Build GRCh38
rs755197118 Chr.22: 49884818-49884818 on Build GRCh38
rs370153801 Chr.22: 49884802-49884802 on Build GRCh38
rs200066649 Chr.22: 49884831-49884831 on Build GRCh38
rs139902588 Chr.22: 49884807-49884807 on Build GRCh38
rs780126311 Chr.22: 49884816-49884816 on Build GRCh38
rs947054618 Chr.22: 49884705-49884705 on Build GRCh38
rs767132038 Chr.22: 49884721-49884721 on Build GRCh38
rs762485782 Chr.22: 49884646-49884646 on Build GRCh38
rs150475592 Chr.22: 49884708-49884708 on Build GRCh38
rs558132731 Chr.22: 49884634-49884634 on Build GRCh38
rs772292224 Chr.22: 49884610-49884610 on Build GRCh38
rs764585141 Chr.22: 49884739-49884739 on Build GRCh38
rs571576115 Chr.22: 49884700-49884700 on Build GRCh38
rs146641467 Chr.22: 49884668-49884668 on Build GRCh38
rs773737919 Chr.22: 49884715-49884715 on Build GRCh38
rs751578085 Chr.22: 49884817-49884817 on Build GRCh38
rs778885980 Chr.22: 49884688-49884688 on Build GRCh38
rs144439900 Chr.22: 49884723-49884723 on Build GRCh38
rs145976876 Chr.22: 49884798-49884798 on Build GRCh38
rs757638894 Chr.22: 49884806-49884806 on Build GRCh38
rs148413052 Chr.22: 49884766-49884766 on Build GRCh38
rs901349363 Chr.22: 49884752-49884752 on Build GRCh38
rs768813845 Chr.22: 49884773-49884773 on Build GRCh38
rs201630531 Chr.22: 49884677-49884677 on Build GRCh38
rs749782154 Chr.22: 49884679-49884679 on Build GRCh38
rs149232276 Chr.22: 49884718-49884718 on Build GRCh38
rs756589477 Chr.22: 49884731-49884731 on Build GRCh38
rs910798 Chr.22: 49884790-49884790 on Build GRCh38
rs533958242 Chr.22: 49884789-49884789 on Build GRCh38
rs761532490 Chr.22: 49884628-49884628 on Build GRCh38
rs781388058 Chr.22: 49884825-49884825 on Build GRCh38
rs6009915 Chr.22: 49884636-49884636 on Build GRCh38
rs369104595 Chr.22: 49884676-49884676 on Build GRCh38
rs765834662 Chr.22: 49884653-49884653 on Build GRCh38
rs570471170 Chr.22: 49884753-49884753 on Build GRCh38
rs143906560 Chr.22: 49884756-49884756 on Build GRCh38
rs138332752 Chr.22: 49884709-49884709 on Build GRCh38
rs1005649197 Chr.22: 49884758-49884758 on Build GRCh38
rs776307325 Chr.22: 49884624-49884624 on Build GRCh38
rs200590187 Chr.22: 49884669-49884669 on Build GRCh38
rs749014874 Chr.22: 49884786-49884786 on Build GRCh38
rs754263155 Chr.22: 49884804-49884804 on Build GRCh38
rs775780599 Chr.22: 49884699-49884699 on Build GRCh38
rs751121432 Chr.22: 49884667-49884667 on Build GRCh38
rs774674482 Chr.22: 49884604-49884604 on Build GRCh38
rs773004333 Chr.22: 49884784-49884784 on Build GRCh38
rs757722107 Chr.22: 49884754-49884754 on Build GRCh38
rs376883523 Chr.22: 49884799-49884799 on Build GRCh38
rs780292820 Chr.22: 49884762-49884762 on Build GRCh38
rs139228464 Chr.22: 49884670-49884670 on Build GRCh38
rs755339556 Chr.22: 49884783-49884783 on Build GRCh38
rs765213568 Chr.22: 49884633-49884633 on Build GRCh38
rs772191419 Chr.22: 49884697-49884697 on Build GRCh38
rs374732866 Chr.22: 49884684-49884684 on Build GRCh38
rs759708957 Chr.22: 49884606-49884606 on Build GRCh38

View Full Product Details