Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010774.2 | NP_034904.2 | 7 | 1649 | |
GenBank mRNA | AF072249.1 | - | 7 | 1666 | |
AK147782.1 | - | 4 | 631 | ||
AK161005.1 | - | 7 | 2637 | ||
BC024812.1 | - | 7 | 1631 | 5356016 |