Gene Transcripts

Gene Symbol
Pax8
Entrez Gene ID
18510
Gene Name
paired box 8
Gene Aliases
Pax-8
Chromosome Location
Chr. 2: 24420551 - 24480092 on Build GRCm38
UniGene ID
Mm.2533
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011040.4 NP_035170.1 11, 12 1497
GenBank mRNA AK050717.1 - 11, 12 1473
AK052610.1 - 11, 12 1474
BC020526.1 - 11, 12 1464 4239835
X57487.1 - 11, 12 1454

View Full Product Details