Gene Transcripts
- Gene Symbol
- Gm3579
- Entrez Gene ID
- 100041932
- Gene Name
- predicted gene 3579
- Gene Aliases
- H3097D08
- Chromosome Location
- Chr. 4: 133095962 - 133101555 on Build GRCm38
- UniGene ID
- Mm.484448
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY140896.1 |
- |
1 |
3812 |
|
BC066867.1 |
- |
1 |
1720 |
30462835 |
- Gene Symbol
- Gm42035
- Entrez Gene ID
- 105246811
- Gene Name
- predicted gene, 42035
- Gene Aliases
- Chromosome Location
- Chr. 8: 90888875 - 90891890 on Build GRCm38
- Species
- Mus musculus
View Full Product Details