Gene Transcripts

Gene Symbol
Diap2
Entrez Gene ID
54004
Gene Name
diaphanous homolog 2 (Drosophila)
Gene Aliases
Dia3, Diaph2, Drf2, E430022I22Rik
Chromosome Location
Chr. X: 129749668 - 130465834 on Build GRCm38
UniGene ID
Mm.10763
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172493.2 NP_766081.1 26 3460
GenBank mRNA AB713187.1 - 27 3255
AK083254.1 - 19 2392
AK088648.1 - 26 3394
AY312280.1 - 26 3397
BB430043.2 - 3 342
BC100759.1 - 3 507
Y15910.1 - 7 967

View Full Product Details