Gene Transcripts

Gene Symbol
Dnajb13
Entrez Gene ID
69387
Gene Name
DnaJ (Hsp40) related, subfamily B, member 13
Gene Aliases
1700014P03Rik, Tsarg
Chromosome Location
Chr. 7: 100503011 - 100514815 on Build GRCm38
UniGene ID
Mm.278554
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153527.2 NP_705755.2 3 350
GenBank mRNA AF419292.2 - 3 495
AK005984.1 - 3 367
AK139680.1 - 322
BC048501.1 - 3 350 6705915

View Full Product Details