Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_175226.4 | NP_780435.1 | 2 | 1463 | |
GenBank mRNA | AK028400.1 | - | 2 | 1416 | |
AK029069.1 | - | 2 | 1466 | ||
AK033506.1 | - | 2 | 1397 | ||
BC052901.1 | - | 2 | 1412 | 30070643 |